missing translation for 'onlineSavingsMsg'
Learn More

PlGF-2 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF465

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488834

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

PlGF-2 Polyclonal antibody AF465 Specifically detects PlGF-2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

PlGF-2
LYOPH
Western Blot 0.1 ug/mL
Q544A5
Pgf
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse PlGF-2 (R&D Systems, Catalog # 465-PL), Ala24-Pro158, Accession # Q544A5
Immunogen affinity purified
RUO
Primary
5281
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse PlGF-2 in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant human PlGF is observed.
Mouse
Videos

For Research Use Only