missing translation for 'onlineSavingsMsg'
Learn More

PILR-beta Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3525

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448214

  • £349.86 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PILR-beta Polyclonal antibody AF3525 Specifically detects PILR-beta in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

PILR-beta
LYOPH
Western Blot 0.1 ug/mL
Q2YFS2
activating receptor PILRbeta, Activating receptor PILR-beta, Cell surface receptor FDFACT, cell surface receptor FDFACT1, cell surface receptor FDFACT2, FDFACT, FDFACT1, FDFACT2, paired immunoglobin-like receptor beta, paired immunoglobin-like type 2 receptor beta, paired immunoglobulin-like receptor beta, paired immunoglobulin-like type 2 receptor beta, PILRB, PILRbeta
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse PILR-beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse PILR-alpha is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Pilrb1
Mouse myeloma cell line NS0-derived recombinant mouse PILR-beta Ala28-Gly193 Accession # Q2YFS2
Immunogen affinity purified
RUO
Primary
29990
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only