missing translation for 'onlineSavingsMsg'
Learn More

PGLYRP1/PGRP-S Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2696

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487724

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

PGLYRP1/PGRP-S Polyclonal antibody AF2696 Specifically detects PGLYRP1/PGRP-S in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

PGLYRP1/PGRP-S
LYOPH
Western Blot 0.1 ug/mL
O88593
peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRPS, PGRP-S, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse PGRP-S in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human PGRP-S is observed and less than 5% cross-reactivity with recombinant mouse PGRP-L is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Pglyrp1
Mouse myeloma cell line NS0-derived recombinant mouse PGLYRP1/PGRP-S, Cys17-Glu182, Accession # O88593
Immunogen affinity purified
RUO
Primary
8993
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only