missing translation for 'onlineSavingsMsg'
Learn More

Persephin Sheep anti-Mouse, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2479

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467584

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Persephin Polyclonal antibody AF2479 Specifically detects Persephin in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Persephin
LYOPH
Western Blot 0.1 ug/mL
O70300.1
persephin, PSP, PSPN
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Persephin in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human Persephin is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PSPN
E. coli-derived recombinant mouse Persephin Ala61-Gly156 Accession # O70300.1
Immunogen affinity purified
RUO
Primary
5623
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only