missing translation for 'onlineSavingsMsg'
Learn More

OX40/TNFRSF4 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1256

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13446844

  • £433.65 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

OX40/TNFRSF4 Polyclonal antibody AF1256 Specifically detects OX40/TNFRSF4 in Mouse samples. It is validated for Agonist Activity, Western Blotting
Specifications

Specifications

OX40/TNFRSF4
LYOPH
Western Blot 0.1 ug/mL, Agonist Activity
P47741
ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LOX40 antigen
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse OX40/TNFRSF4 in direct ELISAs and Western blots. In these formats, less than 2% cross-reactivity with recombinant mouse (rm) EDAR, rm4-BB, rmCD27, rmDR3, rmGITR, rmNGF R, rmCD30, rmCD40, rmFas, rmOPG, rmRANK, rmTNF RI, and rmTNF RII is observed.
Mouse
Western Blot, Agonist Activity
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TNFRSF4
Mouse myeloma cell line NS0-derived recombinant mouse OX40/TNFRSF4 Val20-Pro211 Accession # P47741
Immunogen affinity purified
RUO
Primary
7293
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only