Learn More
Osteoadherin/OSAD Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3308
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488094
  • £382.00 / 100µg
Estimated Shipment
05-07-2018
Add to basket
Description
Description
Osteoadherin/OSAD Polyclonal antibody AF3308 Specifically detects Osteoadherin/OSAD in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Osteoadherin/OSAD
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse Osteoadherin/OSAD, Gln21-Ile423, Accession No. O35103
100μg
Primary
Murine
Unconjugated
O35103
Goat