Learn More
OSM R beta Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF662
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439824
  • £449.92 / 100µg
Estimated Shipment date
from Supplier 10-07-2020
Description
Description
OSMR beta Polyclonal antibody specifically detects OSMR beta in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
OSMR beta
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
O70458
IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSM R beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse OSM R beta in direct ELISAs and Western blots.
Mouse
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Osmr
Mouse myeloma cell line NS0-derived recombinant mouse OSM R beta Glu24-Leu738 Accession # O70458
Immunogen affinity purified
RUO
Primary
9180
Reconstitute at 0.2 mg/mL in sterile PBS.