missing translation for 'onlineSavingsMsg'
Learn More

OCIL/CLEC2d Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3376

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448134

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 24-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

OCIL/CLEC2d Polyclonal antibody AF3376 Specifically detects OCIL/CLEC2d in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

OCIL/CLEC2d
LYOPH
Western Blot 0.1 ug/mL
Q91V08
CLAXLLT-1, CLEC2D, C-type lectin domain family 2 member D, C-type lectin domain family 2, member D, C-type lectin related f, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT1, LLT1Osteoclast inhibitory lectin, OCILmember D
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse OCIL/CLEC2d in direct ELISAs and Western blots. In Western blots, approximately 35% cross-reactivity with recombinant mouse OCILRP2 is observed and less than 2% cross-reactivity with recombinant human OCIL is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
CLEC2D
E. coli-derived recombinant mouse OCIL/CLEC2d Ala65-Ser207 Accession # Q91V08
Immunogen affinity purified
RUO
Primary
29121
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only