missing translation for 'onlineSavingsMsg'
Learn More

OCAM/NCAM2 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF778

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460205

  • £389.41 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

OCAM/NCAM2 Polyclonal antibody AF778 Specifically detects OCAM/NCAM2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

OCAM/NCAM2
LYOPH
Western Blot 0.1 ug/mL
O35136
NCAM2, N-CAM-2, NCAM-2, NCAM21MGC51008, neural cell adhesion molecule 2, neural cell adhesion molecule 2 precursor10, Rb-8, RNCAM
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse OCAM in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) NCAM-1 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NCAM2
Mouse myeloma cell line NS0-derived recombinant mouse OCAM, Leu20-Gly700, Accession # O35136
Antigen Affinity-purified
RUO
Primary
4685
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only