missing translation for 'onlineSavingsMsg'
Learn More

NPR-1 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3974

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438474

  • £289.09 / 100µg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NPR-1 Polyclonal antibody AF3974 Specifically detects NPR-1 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

NPRA/NPR1
LYOPH
Western Blot 0.1 ug/mL
P18293
ANPa, ANP-A, ANPRA, ANPR-A, ANPRAEC 4.6.1.2, atrial natriuretic peptide receptor 1, Atrial natriuretic peptide receptor type A, atrionatriuretic peptide receptor A, GC-A, Guanylate cyclase A, GUC2A, GUCY2A, natriuretic peptide A type receptor, natriuretic peptide receptor A, natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptidereceptor A), NPR1, NPRA, NPR-A, Pndr
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse NPRA/NPR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) NPR1 is observed and 5% cross-reactivity with rhNPR2 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NPR1
Mouse myeloma cell line NS0-derived recombinant mouse NPRA/NPR1 Ser29-Glu469 Accession # P18293
Antigen Affinity-purified
RUO
Primary
4881
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only