missing translation for 'onlineSavingsMsg'
Learn More

NOV/CCN3 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1976

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437264

  • £389.41 / 100µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NOV/CCN3 Polyclonal antibody AF1976 Specifically detects NOV/CCN3 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

NOV/CCN3
LYOPH
Western Blot 0.1 ug/mL
Q64299
CCN family member 3, CCN3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse NOV/CCN3 in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human NOV is observed and less than 1% cross-reactivity with recombinant mouse WISP-1 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NOV
Mouse myeloma cell line NS0-derived recombinant mouse NOV/CCN3 Ser26-Ile354 Accession # Q64299
Immunogen affinity purified
RUO
Primary
4856
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only