missing translation for 'onlineSavingsMsg'
Learn More

Notch-3 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1308

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13456874

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Notch-3 Polyclonal antibody AF1308 Specifically detects Notch-3 in Mouse samples. It is validated for Blocking (Receptor-ligand), Flow Cytometry, Immunohistochemistry, Western Blotting
Specifications

Specifications

Notch-3
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction, CyTOF-ready
Q61982
CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3, Notch3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Notch-3 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human Notch-3 is observed and less than 5% cross-reactivity with recombinant rat (rr) Notch-1 and rrNotch-2 is observed.
Mouse
Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NOTCH3
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Notch-3 Ala40-Glu468 Accession # Q61982
Antigen Affinity-purified
RUO
Primary
4854
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only