Learn More
Neuropoietin/NP Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2709
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13447734
  • £332.22 / 100µg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
Neuropoietin/NP Polyclonal antibody AF2709 Specifically detects Neuropoietin/NP in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Neuropoietin/NP
LYOPH
Western Blot 0.1 ug/mL
P83714
Ctf2
E. coli-derived recombinant mouse Neuropoietin/NP Ala23-Ala204 Accession # P83714
Immunogen affinity purified
RUO
Primary
100289419
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Neuropoietin/NP in direct ELISAs and Western blots.