missing translation for 'onlineSavingsMsg'
Learn More

Netrin-1 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1109

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13446744

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Netrin-1 Polyclonal antibody AF1109 Specifically detects Netrin-1 in Mouse samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications

Specifications

Netrin-1
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
AAC52971
netrin 1, netrin-1, Netrin1, NTN1, NTN1Lnetrin 1, mouse, homolog of
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Netrin-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant chicken (rch) Netrin-1 and recombinant human Netrin-1 is observed and less than 1% cross-reactivity with recombinant mouse (rm) Netrin-G1a and rmNetrin-4 is observed.
Mouse
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NTN1
Mouse myeloma cell line NS0-derived recombinant mouse Netrin-1 Val22-Ala603 Accession # AAC52971
Antigen Affinity-purified
RUO
Primary
9423
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only