Learn More
Nephronectin Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF4298
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488654
  • £382.00 / 100µg
Estimated Shipment
02-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
Nephronectin Polyclonal antibody AF4298 Specifically detects Nephronectin in Mouse samples. It is validated for Immunohistochemistry
Specifications
Specifications
Nephronectin
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse Nephronectin, Asp20-Cys561, Accession No. AAH68308
100μg
Primary
114249
Unconjugated
AAH68308
Goat
IgG
RUO
Polyclonal
Murine
Videos