Learn More
Nectin-4 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3116
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487984
  • £382.00 / 100µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
Nectin-4 Polyclonal antibody AF3116 Specifically detects Nectin-4 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Nectin-4
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse Nectin-4, Tyr28-Ile349, Accession No. Q8R007
100μg
Primary
71740
Unconjugated
Q8R007
Goat
IgG
RUO
Polyclonal
Murine
Videos