missing translation for 'onlineSavingsMsg'
Learn More

Nectin-3 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3498

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438204

  • £349.86 / 100µg

Estimated Shipment date
from Supplier 23-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Nectin-3 Polyclonal antibody AF3498 Specifically detects Nectin-3 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Nectin-3/PVRL3
LYOPH
Western Blot 0.1 ug/mL
Q9JLB9.1
CD113, CD113 antigen, CDw113FLJ90624, DKFZp566B0846, nectin 3, nectin-3, Nectin3, poliovirus receptor-related 3, poliovirus receptor-related protein 3, PPR3, PRR3CD113, PVRL3, PVRR3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Nectin-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human Nectin-3 is observed and less than 2% cross-reactivity with recombinant mouse (rm) Nectin-1 and rmNectin-4 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PVRL3
Mouse myeloma cell line NS0-derived recombinant mouse Nectin-3 Leu56-Asp400 Accession # Q9JLB9.1
Immunogen affinity purified
RUO
Primary
25945
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only