missing translation for 'onlineSavingsMsg'
Learn More

MMP-3 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF548

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13429274

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MMP-3 Polyclonal antibody AF548 Specifically detects MMP-3 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

MMP-3
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
P28862
CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP3, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, Stromelysin 1, stromelysin-1, Transin-1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse MMP-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human MMP-3 is observed and less than 1% cross-reactivity with recombinant mouse MMP-2 is observed.
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MMP3
Mouse myeloma cell line NS0-derived recombinant mouse MMP-3 Tyr18-Cys477 Accession # P28862
Immunogen affinity purified
RUO
Primary
4314
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only