Learn More
Mimecan Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2949
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487884
  • £382.00 / 100µg
Estimated Shipment
01-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
Mimecan Polyclonal antibody AF2949 Specifically detects Mimecan in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Mimecan
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse Mimecan, Ala20-Phe298, Accession No. Q62000
100μg
Primary
18295
Unconjugated
Q62000
Goat
IgG
RUO
Polyclonal
Murine
Videos