missing translation for 'onlineSavingsMsg'
Learn More

MGL2/CD301b Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2835

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467814

  • £349.86 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MGL2/CD301b Polyclonal antibody AF2835 Specifically detects MGL2/CD301b in Mouse samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications

Specifications

MGL2/CD301b
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
NP_660119
Mgl2
Mouse myeloma cell line NS0-derived recombinant mouse MGL2/CD301b, Ser72-Pro332, Accession # NP_660119
Immunogen affinity purified
RUO
Primary
10462
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse MGL2/CD301b in direct ELISAs and Western blots.
Mouse
Videos
Documents

For Research Use Only