missing translation for 'onlineSavingsMsg'
Learn More

MFRP Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3445

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428174

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MFRP Polyclonal antibody AF3445 Specifically detects MFRP in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

MFRP
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Q8K480
MCOP5, membrane frizzled-related protein, Membrane-type frizzled-related protein, MGC32938, NNO2, RD6
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse MFRP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human (rh) MFRP is observed, 15% cross-reactivity with recombinant mouse (rm) Frizzled-4 is observed, 10% cross-reactivity with rmFrizzled-1, rhFrizzled-5, rmFrizzled-8, and rmFrizzled-10 is observed, and 5% cross-reactivity with rmFrizzled-3, rmFrizzled-6, and rmFrizzled-7 is observed.
Mouse
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MFRP
Mouse myeloma cell line NS0-derived recombinant mouse MFRP Gln91-Pro584 Accession # Q8K480
Immunogen affinity purified
RUO
Primary
83552
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only