missing translation for 'onlineSavingsMsg'
Learn More

Meprin alpha Subunit/MEP1A Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4007

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13468494

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Meprin alpha Subunit/MEP1A Polyclonal antibody AF4007 Specifically detects Meprin alpha Subunit/MEP1A in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Meprin alpha Subunit/MEP1A
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
NP_032611
bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, MEP1A, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Meprin alpha Subunit/MEP1A in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant human MEP1A is observed and less than 5% cross-reactivity with recombinant mouse MEP1B is observed.
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MEP1A
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Meprin alpha Subunit/MEP1A Val34-Arg615 Accession # NP_032611
Antigen Affinity-purified
RUO
Primary
4224
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only