Learn More
MEA-1 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF3088
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13437974
  • £289.09 / 100µg
Estimated Shipment
08-07-2019
Description
Description
MEA-1 Polyclonal antibody AF3088 Specifically detects MEA-1 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
MEA-1
LYOPH
Western Blot 0.1 ug/mL
Q64327
HYS, male-enhanced antigen, Male-enhanced antigen (H-Y structural gene), male-enhanced antigen 1, MEA1, MEAMEA-1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse MEA-1 in direct ELISAs and Western blots.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MEA1
E. coli-derived recombinant mouse MEA-1 Ala2-Lys174 Accession # Q64327
Immunogen affinity purified
RUO
Primary
4201
Reconstitute at 0.2 mg/mL in sterile PBS.