missing translation for 'onlineSavingsMsg'
Learn More

Matrilin-4 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3380

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488134

  • £357.73 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Matrilin-4 Polyclonal antibody AF3380 Specifically detects Matrilin-4 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Matrilin-4
LYOPH
Western Blot 0.1 ug/mL
O89029
FLJ14417, MATN4, matrilin 4, matrilin-4, Matrilin4
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse Matrilin-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) Matrilin-4 is observed and less than 2% cross-reactivity with recombinant mouse Matrilin-2 and rhMatrilin-3 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MATN4
Mouse myeloma cell line NS0-derived recombinant mouse Matrilin-4, Gln22-Lys624, Accession # O89029
Immunogen affinity purified
RUO
Primary
8785
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only