missing translation for 'onlineSavingsMsg'
Learn More

Lipin 3 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4064

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488524

  • £347.17 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

Lipin 3 Polyclonal antibody AF4064 Specifically detects Lipin 3 in Mouse samples. It is validated for Immunocytochemistry, Western Blotting
Specifications

Specifications

Lipin 3
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
E. coli-derived recombinant mouse Lipin 3 , Thr428-Arg529, Accession No. Q99PI4
100μg
Primary
64899
Unconjugated
Q99PI4
Goat
IgG
RUO
Polyclonal
Murine
Videos

For Research Use Only