Learn More
LIGHT/TNFSF14 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1794
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487154
  • £382.00 / 100µg
Estimated Shipment
05-07-2018
Add to basket
Description
Description
LIGHT/TNFSF14 Polyclonal antibody AF1794 Specifically detects LIGHT/TNFSF14 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
LIGHT/TNFSF14
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse LIGHT, Asp72-Val239, Accession No. Q9QYH9
100μg
Primary
50930
Unconjugated
Q9QYH9
Goat
IgG
RUO
Polyclonal
Murine