missing translation for 'onlineSavingsMsg'
Learn More

Layilin Ectodomain Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2646

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467694

  • £342.51 / 100µg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Layilin Ectodomain Polyclonal antibody AF2646 Specifically detects Layilin Ectodomain in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Layilin
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q8C351
FLJ30977, FLJ31092, layilin, LAYN, LYAN
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Layilin Ectodomain in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant human Layilin is observed.
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
LAYN
Mouse myeloma cell line NS0-derived recombinant mouse Layilin Ectodomain Lys18-Glu229 (Ala29-Gly36 del) Accession # Q8C351
Immunogen affinity purified
RUO
Primary
143903
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only