missing translation for 'onlineSavingsMsg'
Learn More

Latent TGF-beta bp4 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2885

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467844

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Latent TGF-beta bp4 Polyclonal antibody AF2885 Specifically detects Latent TGF-beta bp4 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Latent TGF-beta bp4
LYOPH
Western Blot 0.1 ug/mL
AAI41006
Latent TGFbeta bp4, latent transforming growth factor beta binding protein 4, LTBP4, LTBP-4, LTBP4L, LTBP4S
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Latent TGF-beta bp4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) LTBP-1 and rhLTBP-2 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
LTBP4
Mouse myeloma cell line NS0-derived recombinant mouse Latent TGF-beta bp4 isoform 1 Ser25-Ala1666 (Arg542-Pro547) Accession # AAI41006
Immunogen affinity purified
RUO
Primary
8425
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only