missing translation for 'onlineSavingsMsg'
Learn More

L-Selectin/CD62L Chicken anti-Mouse, Polyclonal, R&D Systems™

Chicken Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF576

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13469414

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

L-Selectin/CD62L Polyclonal antibody AF576 Specifically detects L-Selectin/CD62L in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

L-Selectin/CD62L
LYOPH
Western Blot 0.1 ug/mL
P18337
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM1, LAM-1, LAM1LECAM1, LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHR, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L, SELL, TQ1
Chicken
IgY
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse L-Selectin/CD62L in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human L-Selectin is observed and less than 2% cross-reactivity with recombinant mouse P-Selectin is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SELL
Mouse myeloma cell line NS0-derived recombinant mouse L-Selectin/CD62L Met1-Asn332 Accession # P18337
Antigen Affinity-purified from egg yolks
RUO
Primary
6402
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only