Learn More
JAM-A Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1077
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13486714
  • £377.53 / 100µg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
JAM-A Polyclonal antibody AF1077 Specifically detects JAM-A in Mouse samples. It is validated for ELISA Capture, Immunohistochemistry, Western Blotting
Specifications
Specifications
JAM-A
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse JAM-A, Lys27-Ala242, Accession No. O88792
100μg
Primary
16456
Unconjugated
O88792
Goat
IgG
RUO
Polyclonal
Murine