missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta 6 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2389

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487524

  • £377.53 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Integrin beta 6 Polyclonal antibody AF2389 Specifically detects Integrin beta 6 in Mouse samples. It is validated for Immunocytochemistry, Western Blotting
Specifications

Specifications

Integrin beta 6
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin β6, Gly22-Asn706, Accession No. Q9Z0T9
100μg
Primary
16420
Unconjugated
Q9Z0T9
Goat
IgG
RUO
Polyclonal
Murine
Videos

For Research Use Only