Learn More
Integrin alpha E/CD103 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1990
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13437274
  • £433.65 / 100µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Integrin alpha E/CD103 Polyclonal antibody AF1990 Specifically detects Integrin alpha E/CD103 in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Specifications
Specifications
Integrin alpha E/CD103
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q60677
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), ITGAE, MGC141996, Mucosal lymphocyte 1 antigen
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Integrin alpha E/CD103 in direct ELISAs and Western blots.
Mouse
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ITGAE
Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha E/CD103 Phe20-Arg1113 Accession # Q60677
Antigen Affinity-purified
RUO
Primary
3682
Reconstitute at 0.2 mg/mL in sterile PBS.