missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 3/CD49c Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2787

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487784

  • £382.00 / 100µg


Estimated Shipment
07-03-2018
Add to basket

Disclaimers:

For Research Use Only


Description

Description

Integrin alpha 3/CD49c Polyclonal antibody AF2787 Specifically detects Integrin alpha 3/CD49c in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

Integrin alpha 3/CD49c
Unconjugated
Q62470
Goat
IgG
RUO
Polyclonal
Detects mouse Integrin alpha 3/CD49c in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) Integrin alpha 3 is observed.
Western Blotting
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Affinity Purified
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Integrin α3/CD49c, Phe33-Glu993, Accession No. Q62470
100μg
Primary
16400
Murine
Videos