missing translation for 'onlineSavingsMsg'
Learn More

IL-7 R alpha/CD127 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF747

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440125

  • £390.01 / 100µg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-7R alpha/CD127 Polyclonal antibody specifically detects IL-7R alpha/CD127 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF.
Specifications

Specifications

IL-7R alpha/CD127
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q9R0C1
CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse IL-7 R alpha in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human IL-7 R alpha is observed.
Mouse
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL7R
Mouse myeloma cell line NS0-derived recombinant mouse IL-7 R alpha/CD127 Glu21-Asp239 Accession # Q9R0C1
Immunogen affinity purified
RUO
Primary
3575
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only