Learn More
anti-Mouse IL-7, Polyclonal, R&D Systems™
Brand:  RD Systems (Bio-Techne) AB-407-NA
Code : Z1
Additional Details : Weight : 0.00010kg
Product Code. 13486604
  • £382.00 / 1mg
Estimated Shipment
02-07-2018
Add to basket
Specifications
Specifications
IL-7
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
E. coli-derived recombinant mouse IL-7, Glu26-Ile154, Accession No. Q544C8
1mg
Primary
16196
Unconjugated
Q544C8
Goat
IgG
RUO
Polyclonal
Murine
Videos