missing translation for 'onlineSavingsMsg'
Learn More

IL-5 R alpha/CD125 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF553

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459294

  • £384.13 / 100µg


Estimated Shipment
29-11-2018
Add to basket

Description

Description

IL-5 R alpha/CD125 Polyclonal antibody AF553 Specifically detects IL-5 R alpha/CD125 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

IL-5 R alpha/CD125
LYOPH
Western Blot 0.1 ug/mL
P21183
CD125, CD125 antigen, CDw125CD125, HSIL5R3, IL-5 receptor subunit alpha, IL5R, IL5R alpha, IL-5R subunit alpha, IL5RA, IL-5RA, IL-5R-alpha, interleukin 5 receptor type 3, interleukin 5 receptor, alpha, interleukin-5 receptor alpha chain, interleukin-5 receptor alpha subunit, interleukin-5 receptor subunit alpha, MGC26560
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects mouse IL-5 R alpha/CD125 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) IL-5 R alpha is observed and less than 1% cross-reactivity with recombinant mouse (rm) IL-4 R, rhIL-9 R, rhIL-13 R alpha 1, and rhIL-13 R alpha 2 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL5RA
Mouse myeloma cell line NS0-derived recombinant mouse IL-5 R alpha/CD125, Asp18-His339, Accession # P21183
Antigen Affinity-purified
RUO
Primary
3568
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only