missing translation for 'onlineSavingsMsg'
Learn More

IL-4 R alpha Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF530

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13439184

  • £389.41 / 100µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-4R alpha Polyclonal antibody specifically detects IL-4R alpha in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF.
Specifications

Specifications

IL-4R alpha
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Affinity Purified
IL4RA
Mouse myeloma cell line NS0-derived recombinant mouse IL-4 R alpha
Immunogen affinity purified
RUO
Primary
3566
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD124, CD124 antigen, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse IL-4 R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) IL-4 R, rhIL-9 R, rhIL-13 R alpha 1, and recombinant mouse IL-13 R alpha 2 is observed.
Mouse
Videos

For Research Use Only