missing translation for 'onlineSavingsMsg'
Learn More

IL-31 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3028

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447934

  • £358.68 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-31 Polyclonal antibody AF3028 Specifically detects IL-31 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

IL-31
LYOPH
Western Blot 0.1 ug/mL
Q6EAL8
IL31, IL-31interleukin-31, interleukin 31
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse IL-31 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human IL-31 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL31
E. coli-derived recombinant mouse IL-31 Thr24-Cys163 Accession # Q6EAL8
Immunogen affinity purified
RUO
Primary
386653
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only