missing translation for 'onlineSavingsMsg'
Learn More

IL-3 R alpha/CD123 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF983

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460375

  • £304.93 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

IL-3 R alpha/CD123 Polyclonal antibody AF983 Specifically detects IL-3 R alpha/CD123 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

IL-3 R alpha/CD123
LYOPH
Western Blot 0.1 ug/mL
P26952
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL3R alpha, IL-3R subunit alpha, IL3RA, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse IL-3 R alpha/CD123 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human IL-3 R alpha and recombinant mouse IL-3 R beta is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL3RA
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-3 R alpha/CD123 Ser17-Lys331 Accession # P26952
Immunogen affinity purified
RUO
Primary
3563
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only