missing translation for 'onlineSavingsMsg'
Learn More

IL-22 R alpha 1 Sheep anti-Mouse, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4294

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458654

  • £314.17 / 100µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

IL-22 R alpha 1 Polyclonal antibody AF4294 Specifically detects IL-22 R alpha 1 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

IL-22 R alpha 1
LYOPH
Western Blot 0.1 ug/mL
Q80XZ4
CRF2-9, CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R alpha 1, IL22R1, IL22RA1, IL-22RA1, IL-22R-alpha-1, IL-TIF-R1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse IL-22 R alpha 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human IL-22 R is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL22RA1
Mouse myeloma cell line NS0-derived recombinant mouse IL-22 R alpha 1 Thr18-Ala228 Accession # Q80XZ4
Immunogen affinity purified
RUO
Primary
58985
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only