missing translation for 'onlineSavingsMsg'
Learn More

IL-20 R alpha Sheep anti-Mouse, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1736

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447124

  • £342.51 / 100µg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-20R alpha Polyclonal antibody specifically detects IL-20R alpha in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

IL-20R alpha
LYOPH
Western Blot 0.1 ug/mL
Q6PHB0
class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20 R alpha, IL20R alpha, IL-20R1IL-20 receptor subunit alpha, IL20RA, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse IL-20 R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) IL-20 R alpha is observed and less than 5% cross-reactivity with rhIL-20 R beta , rhIFN-gamma RI, rhIFN-gamma RII, and rhIL-10 R beta is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Il22ra2
Mouse myeloma cell line NS0-derived recombinant mouse IL-20 R alpha Val33-Lys253 Accession # Q6PHB0
Antigen Affinity-purified
RUO
Primary
53832
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only