missing translation for 'onlineSavingsMsg'
Learn More

IL-2 R beta Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF589

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449484

  • £349.86 / 100µg

Estimated Shipment date
from Supplier 19-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-2 R beta Polyclonal antibody AF589 Specifically detects IL-2 R beta in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

IL-2 R beta
LYOPH
Western Blot 0.1 ug/mL
P16297
CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-15 R beta, IL-2 receptor subunit beta, IL2R beta, IL-2R subunit beta, IL2RB, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse IL-2 R beta in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human IL-2 R beta is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL2RB
Mouse myeloma cell line NS0-derived recombinant mouse IL-2 R beta Ala27-Glu240 Accession # P16297
Antigen Affinity-purified
RUO
Primary
3560
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only