missing translation for 'onlineSavingsMsg'
Learn More

IL-17E/IL-25 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1399

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13436924

  • £371.48 / 100µg

Estimated Shipment
13-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-17E/IL-25 Polyclonal antibody AF1399 Specifically detects IL-17E/IL-25 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

IL-17E/IL-25
LYOPH
Western Blot 0.1 ug/mL
NP_542767
IL17E, IL-17E, IL17Einterleukin-17E, IL25, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse IL-17E/IL-25 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) IL-17E and recombinant rat IL-17E is observed, and less than 1% cross-reactivity with recombinant mouse (rm) IL-17, rmIL-17B, rhIL-17C, rhIL-17D, and rhIL-17F is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IL25
E. coli-derived recombinant mouse IL-17E/IL-25 Val17-Ala169 Accession # NP_542767
Immunogen affinity purified
RUO
Primary
64806
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only