missing translation for 'onlineSavingsMsg'
Learn More

IL-12 R beta 1 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1998

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487274

  • £352.00 / 100µg


Estimated Shipment
08-03-2018
Add to basket

Description

Description

IL-12 R beta 1 Polyclonal antibody AF1998 Specifically detects IL-12 R beta 1 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

IL-12 R beta 1
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-12 Rβ1, Val32-Glu561, Accession No. Q60837
100μg
Primary
16161
Unconjugated
Q60837
Goat
IgG
RUO
Polyclonal
Murine
Videos

For Research Use Only