Learn More
IGSF4A/SynCAM1 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1459
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13456954
  • £342.51 / 100µg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
IGSF4A/SynCAM1 Polyclonal antibody AF1459 Specifically detects IGSF4A/SynCAM1 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
IGSF4A/SynCAM1/CADM1
LYOPH
Western Blot 0.1 ug/mL
NP_061240
BL2, Cadm1, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, SynCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse IGSF4A/SynCAM1 in direct ELISAs and Western blots.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Ikbkg
Mouse myeloma cell line NS0-derived recombinant mouse IGSF4A/SynCAM1 Ile42-Asp376 Accession # NP_061240
Immunogen affinity purified
RUO
Primary
23705
Reconstitute at 0.2 mg/mL in sterile PBS.