Learn More
Mouse IgG2b Isotype Control, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen PA533227
Code : NEW
Additional Details : Weight : 0.01000kg
Description
PA5-33227 is a mouse IgG2b monoclonal antibody arising from a spontaneous mouse plasmacytoma. The antibody does not react with any human cell surface markers, and may therefore be used as an isotype matched negative control reagent to assess the level of non-specific binding of mouse IgG2b monoclonal antibodies in human cells. PA5-33227 is also suitable for use as a negative control with bovine, sheep, pig, canine, rat and guinea-pig tissues.
Mouse IgG2b Control antibody specifically detects Mouse IgG2b in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmune Assays (RIA).Specifications
Mouse IgG2b | |
1 mg/mL | |
PBS with no preservative | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmune Assays (RIA) | |
Unconjugated | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Mouse IgG2b | |
1 mg | |
Isotype Control (Primary) | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |