missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG1, FITC, PE, PE-Cyanine5, Isotype Control, Invitrogen™

Isotype Control Cocktail

Brand:  Invitrogen SA112475

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11575010

  • £587.56 / 100 tests


Estimated Shipment
23-05-2018
Add to basket

Description

Description

This antibody contains a mix of IgG1 (FITC), IgG1 (RPE) and IgG1 (RPE-Cy5) isotypes.SA1-12475 has been successfully used in FACS procedures to assess non-specific binding of mouse monoclonal antibodies in human samples.
Specifications

Specifications

Mouse IgG1 Tri-Color
Cocktail
PBS with 0.2% carrier protein and 0.08% sodium azide
IgG1
White blood cells.
Affinity chromatography, Affinity chromatography, Affinity chromatography
Store at 4° C, do not freeze
Flow Cytometry
FITC/PE/PE-Cyanine5
Purified
Murine
IgG1
100 Tests
Isotype Control
Videos