missing translation for 'onlineSavingsMsg'
Learn More

IGFBP-2 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF797

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13430225

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IGFBP-2 Polyclonal antibody AF797 Specifically detects IGFBP-2 in Mouse samples. It is validated for Neutralization, Western Blotting
Specifications

Specifications

IGFBP-2
LYOPH
Western Blot 0.1 ug/mL, Neutralization 30 ug/mL
CAA57270
BP2, IBP2, IBP-2, IGF-binding protein 2, IGFBP2, IGFBP-2, IGF-BP53, insulin-like growth factor binding protein 2 (36kD), insulin-like growth factor binding protein 2, 36kDa, insulin-like growth factor-binding protein 2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse IGFBP-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human (rh) IGFBP-2 is observed and less than 1% cross-reactivity with rhIGFBP-1, rhIGFBP-3, rhIGFBP-4, rhIGFBP-5, recombinant mouse (rm) IGFBP-3, rmIGFBP-5, and rmIGFBP-6 is observed.
Mouse
Western Blot, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
IGFBP2
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-2 Glu35-Gln305 Accession # CAA57270
Immunogen affinity purified
RUO
Primary
3485
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only