Learn More
HSF4 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF4088
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488544
  • £382.00 / 100µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
HSF4 Polyclonal antibody AF4088 Specifically detects HSF4 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
HSF4
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
E. coli-derived recombinant mouse HSF4 , Gly343-Pro492, Accession No. Q9R0L1
100μg
Primary
26386
Unconjugated
Q9R0L1
Goat
IgG
RUO
Polyclonal
Murine
Videos