Learn More
Hip Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1568
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13457014
  • £362.60 / 100µg
Estimated Shipment
14-09-2020
Description
Description
Hip Polyclonal antibody AF1568 Specifically detects Hip in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Hip
LYOPH
Western Blot 0.1 ug/mL
Q7TN16
FLJ90230, hedgehog interacting protein, hedgehog-interacting protein, HHIP, HIPFLJ20992
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse Hip in direct ELISAs and Western blots.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
HHIP
Mouse myeloma cell line NS0-derived recombinant mouse Hip Lys24-Arg678 (Asn29Ser) Accession # Q7TN16
Immunogen affinity purified
RUO
Primary
64399
Reconstitute at 0.2 mg/mL in sterile PBS.